missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-KIR2DL1/CD158a, PE, Clone: 143211, R&D Systems™
Mouse Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems FAB1844P-025
This item is not returnable.
View return policy
Description
KIR2DL1/KIR2DS5 Monoclonal specifically detects KIR2DL1/KIR2DS5 in Human samples. It is validated for Flow Cytometry.Specifications
| KIR2DL1/KIR2DS5 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD158a, CD158g, NKAT9 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3802 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 143211 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P43626 | |
| KIR2DL1 | |
| BaF3 mouse pro-B cell line transfected with human KIR2DL1/KIR2DS5 Accession # P43626 | |
| 25 Tests | |
| Primary | |
| Detects human KIR2DL1/KIR2DS5 on transfected cells. It does not stain cells transfected with KIR2DL2, 2DL3, 2DL4, 2DL5, 2DS1, 2DS2, 2DS4, 3DL1, 3DL2, or 3DS1. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?