missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kruppel-like factor 2 (lung), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant KLF2.
Brand: Abnova H00010365-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: MALSEPILPSFSTFASPCRERGLQERWPRAEPESGGTDDDLNSVLDFILSMGLDGLSpecifications
| Kruppel-like factor 2 (lung) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant KLF2. | |
| KLF2 | |
| LKLF | |
| Mouse | |
| 50 μL | |
| Transcription Regulation | |
| Primary | |
| Human, Mouse | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_016270 | |
| KLF2 | |
| KLF2 (NP_057354, 1 a.a. ∼ 56 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 10365 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction