missing translation for 'onlineSavingsMsg'
Learn More
Learn More
kynurenine 3-monooxygenase (kynurenine 3-hydroxylase), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human KMO protein.
Brand: Abnova H00008564-B01P.50ug
This item is not returnable.
View return policy
Description
Kynurenine 3-monooxygenase (KMO; EC 1.14.13.9) is an NADPH-dependent flavin monooxygenase that catalyzes the hydroxylation of the L-tryptophan metabolite L-kynurenine to form L-3-hydroxykynurenine.[supplied by OMIM
Sequence: MDSSVIQRKKVAVIGGGLVGSLQACFLAKRNFQIDVYEAREDTRVATFTRGRSINLALSHRGRQALKAVGLEDQIVSQGIPMRARMIHSLSGKKSAIPYGTKSQYILSVSRENLNKDLLTAAEKYPNVKMHFNHRLLKCNPEEGMITVLGSDKVPKDVTCDLIVGCDGAYSTVRSHLMKKPRFDYSQQYIPHGYMELTIPPKNGDYAMEPNYLHIWPRNTFMMIALPNMNKSFTCTLFMPFEEFEKLLTSNDVVDFFQKYFPDAIPLIGEKLLVQDFFLLPAQPMISVKCSSFHFKSHCVLLGDAAHAIVPFFGQGMNAGFEDCLVFDELMDKFSNDLSLCLPVFSRLRIPDDHAISDLSMYNYIEKNMERFLHAIMPSTFIPLYTMVTFSRIRYHEAVQRWHWQKRSpecifications
| kynurenine 3-monooxygenase (kynurenine 3-hydroxylase) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human KMO protein. | |
| KMO | |
| dJ317G22.1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 8564 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC005297.1 | |
| KMO | |
| KMO (AAH05297.1, 1 a.a. ∼ 407 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction