missing translation for 'onlineSavingsMsg'
Learn More
Learn More
kinase suppressor of ras 2, Mouse, Clone: 6C9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant KSR2.
Brand: Abnova H00283455-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: PAPPLPPSATPPSPLHPSPQCTRQQKNFNLPASHYYKYKQQFIFPDVVPVPETPTRAPQVILHPVTSNPILEGNPLLQIEVEPTSENEEVSpecifications
| kinase suppressor of ras 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_173598 | |
| KSR2 | |
| KSR2 (NP_775869, 411 a.a. ∼ 500 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Kinases | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 6C9 | |
| Mouse monoclonal antibody raised against a partial recombinant KSR2. | |
| KSR2 | |
| FLJ25965 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 283455 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction