Learn More
L1 Protein of Human Papilloma Virus Mouse anti-Human, Unlabeled, Clone: CAMVIR-1, BD
Description
More than 60 different types of human papilloma viruses (HPVs) have been isolated. The human papilloma viruses encode three late proteins, which are produced only in terminally differentiating keratinocytes, two of which (the L1 and L2 proteins) are structural components of the virion. Virus-like particles can be assembled by over-expressing L1 and L2 in vitro, demonstrating that L1 and L2 are necessary and sufficient components of the HPV capsomeres. HPV-16 migrates at a reduced molecular weight of ∽57kda on SDS/PAGE. The CAMVIR-1 antibody reacts with the L1 protein of HPV-16. It may cross-react with HPV-33, but does not cross-react with HPV-6 or HPV-11.
Host Species: Mouse
Clone: CAMVIR-1
Isotype: IgG2a κ
Species Reactivity: Viral-transformed cells
Immunofluorescence, Immunohistochemistry, Western Blotting
Specifications
Specifications
| Antigen | L1 Protein of Human Papilloma Virus |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | CAMVIR-1 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.