Learn More
LanC lantibiotic synthetase component C-like 1 (bacterial), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant LANCL1.
Brand: Abnova H00010314-A01.50uL
Description
This gene encodes a loosely associated peripheral membrane protein related to the LanC family of bacterial membrane-associated proteins involved in the biosynthesis of antimicrobial peptides. This protein may play a role as a peptide-modifying enzyme component in eukaryotic cells. Previously considered a member of the G-protein-coupled receptor superfamily, this protein is now in the LanC family. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq
Sequence: MAQRAFPNPYADYNKSLAEGYFDAAGRLTPEFSQRLTNKIRELLQQMERGLKSADPRDSpecifications
| LanC lantibiotic synthetase component C-like 1 (bacterial) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant LANCL1. | |
| LANCL1 | |
| GPR69A/p40 | |
| Mouse | |
| 50 μL | |
| Membrane Receptors | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_006055 | |
| LANCL1 | |
| LANCL1 (NP_006046, 1 a.a. ∼ 58 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 10314 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.