missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN28, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human LIN28 protein.
Brand: Abnova H00079727-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MGSVSNQQFAGGCAKAAEEAPEEAPEDAARAADEPQLLHGAGICKWFNVRMGFGFLSMTARAGVALDPPVDVFVHQSKLHMEGFRSLKEGEAVEFTFKKSAKGLESIRVTGPGGVFCIGSERRPKGKSMQKRRSKGDRCYNCGGLDHHAKECKLPPQPKKCHFCQSISHMVASCPLKAQQGPSAQGKPTYFREEEEEIHSPTLLPEAQNSpecifications
| LIN28 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human LIN28 protein. | |
| LIN28 | |
| CSDD1/FLJ12457/LIN-28/LIN28A/ZCCHC1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79727 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC028566 | |
| LIN28 | |
| LIN28 (AAH28566, 1 a.a. ∼ 209 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction