Learn More
LIM domain only 2 (rhombotin-like 1), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human LMO2 protein.
Brand: Abnova H00004005-D01P.100ug
Description
LMO2 encodes a cysteine-rich, two LIM-domain protein that is required for yolk sac erythropoiesis. The LMO2 protein has a central and crucial role in hematopoietic development and is highly conserved. The LMO2 transcription start site is located approximately 25kb downstream from the 11p13 T-cell translocation cluster (11p13 ttc), where a number T-cell acute lymphoblastic leukemia-specific translocations occur. Alternative splicing results in multiple transcript variants encoding different isoforms
Sequence: MSSAIERKSLDPSEEPVDEVLQIPPSLLTCGGCQQNIGDRYFLKAIDQYWHEDCLSCDLCGCRLGEVGRRLYYKLGRKLCRRDYLRLFGQDGLCASCDKRIRAYEMTMRVKDKVYHLECFKCAACQKHFCVGDRYLLINSDIVCEQDIYEWTKINGMISpecifications
| LIM domain only 2 (rhombotin-like 1) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human LMO2 protein. | |
| LMO2 | |
| RBTN2/RBTNL1/RHOM2/TTG2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4005 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005574.2 | |
| LMO2 | |
| LMO2 (NP_005565.1, 1 a.a. ∼ 158 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.