missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LON peptidase N-terminal domain and ring finger 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human LONRF2 protein.
Brand: Abnova H00164832-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MKNYEQALQDASAACQNEPLLIKGHQVKAQALSGLGRSKEVLKEFLYCLALNPECNSVKKEAQKVMCEVLFSATANVHENLTSSIQSRLKAQGHSHMNAQALLEEGDAGSSENSSEKSDMLGNTNSSVLYFILGLHFEEDKKALESILPTAPSAGLKRQFPDDVEDAPDLNAPGKIPKKDLSPQRSPNSETEESQGLSLDVTDFECALCMRLLFEPVTTPCGHTFCLKCLERCLDHAPHCPLCKDKLSELLASRNFNITVLAEELIFRYLPDELSDRKRIYDEEMSELSNLTRDVPIFVCAMAFPTVPCPLHVFEPRYRLMIRRCMETGTKRFGMCLSAEHAGLSEYGCMLEIKDVRTFPDGSSVVDAIGISRFRVLSHRHRDGYNTADIEYLEDEKVEGPEYEELAALHDSVHQQSVSWFASLQDRMKEQILSHFGVMPDREPEPQSNPSGPAWSWWILAVLPLERKAQLAILGMTSLKERLLAIRRILVIITRKMNSRQELANARERNNSpecifications
| LON peptidase N-terminal domain and ring finger 2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human LONRF2 protein. | |
| LONRF2 | |
| FLJ45273/MGC126711/MGC126713/RNF192 | |
| Mouse | |
| 50 μL | |
| Ubiquitin/Proteasome | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| BC101664.1 | |
| LONRF2 | |
| LONRF2 (AAI01665.1, 1 a.a. ∼ 511 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 164832 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction