Learn More
lymphocyte-specific protein 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human LSP1 protein.
Brand: Abnova H00004046-D01P.100ug
Description
This gene encodes an intracellular F-actin binding protein. The protein is expressed in lymphocytes, neutrophils, macrophages, and endothelium and may regulate neutrophil motility, adhesion to fibrinogen matrix proteins, and transendothelial migration. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Sequence: MAEASSDPGAEEREELLGPTAQWSVEDEEEAVHEQCQHERDRQLQAQDEEGGGHVPERPKQEMLLSLKPSEAPELDEDEGFGDWSQRPEQRQQHEGAQGTLDSGEPPQCRSPEGEQEDRPGLHAYEKEDSDEVHLEELSLSKEGPGPEDTVQDNLGAAGAEEEQEEHQKCQQPRTPSPLVLEGTIEQSSPPLSPTTKLIDRTESLNRSIEKSNSVKKSQPDLPISKIDQWLEQYTQAIETAGRTPKLARQASIELPSMAVASTKSRWETGEVQAQSAAKTPSCKDIVAGDMSKKSLWEQKGGSKTSSTIKSTPSGKRYKFVATGHGKYEKVLVEGGPAPSpecifications
| lymphocyte-specific protein 1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human LSP1 protein. | |
| LSP1 | |
| WP34/pp52 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4046 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC001785.1 | |
| LSP1 | |
| LSP1 (AAH01785.1, 1 a.a. ∼ 339 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.