Learn More
Ly-108 Mouse anti-Mouse, PE, Clone: 13G3, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561540
Description
The 13G3 monoclonal antibody specifically binds to Lymphocyte antigen 108, Ly-108. Ly-108 is a member of the signaling lymphocytic activation molecule (SLAM) family of immune receptors. Ly-108 is a type 1 transmembrane glycoprotein adhesion receptor that is expressed by T cells, NKT cells, B cells, NK cells, macrophages, dendritic cells and neutrophils. Ly-108 plays multiple roles in innate and adaptive immunity including costimulation of NK cell cytotoxicity and T cell cytokine responses. Moreover, Ly-108 has been implicated in autoimmunity.
Specifications
| Ly-108 | |
| Monoclonal | |
| 0.2mg/mL | |
| Slamf6; SLAM family member 6; KAL1; NTB-A; SF2000 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| 13G3 | |
| PE | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| WT thymocytes | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.