Learn More
Ly-6C Rat anti-Mouse, Biotin, Clone: AL-21, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 557359
Description
The AL-21 antibody reacts with a non-polymorphic determinant of Ly-6C, a 14- 17kDa GPI-linked cell-surface antigen found on some monocyte/macrophage populations, granulocytes, endothelial cells, plasma cells, and thymocyte, NK-cell, and T-subsets. Mice with the Ly-6.2 alloantigen (eg, AKR, C57BL, C57BR, C57L, C58, DBA/2, PL, SJL, SWR, 129) have subsets of CD8+ and CD4+ Ly-6C+ T cells, while Ly-6.1 strains (eg, A, BALB/c, CBA, C3H/He, DBA/1, NZB) have only CD8+ Ly-6C+ T cells. Upregulation of Ly-6C expression on CD8+ T cells by interferons α and β and poly (I:C) has been described, and Ly-6C is a memory marker on CD8+ T cells.
Host Species: Rat
Clone: AL-21
Isotype: IgM, κ
Cross Reactivity: Mouse
Flow Cytometry, Immunohistochemistry (Frozen)
Specifications
| Ly-6C | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Not reported | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgM κ |
| Flow Cytometry | |
| AL-21 | |
| Biotin | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.