Learn More
Ly-6D Rat anti-Mouse, Unlabeled, Clone: 49-H4, BD
Description
The 49-H4 antibody reacts with the ThB antigen (Ly-6D), which is a 15kDa GPI-anchored protein of the Ly-6 multigene family. ThB has been detected on cortical thymocytes, small sIg- Thy-1- and sIg+ lymphoid cells in the bone marrow, thymic medullary epithelial cells, all epidermal layers (except the stratum corneum), and on peripheral B lymphocytes, but not on peripheral T lymphocytes. It has been noted that there is strain-to-strain variation in the expression of the ThB antigen on splenic B cells, those of strains with the Ly-6.2 haplotype (e.g., AKR, C57BL, DBA/2, SJL, SWR) stain more intensely with anti- ThB reagents than those of Ly-6.1 strains (e.g., A, BALB/c, CBA, C3H/He, NZB), and B cells of hybrids of Ly-6.1 and Ly-6.2 strains stain with intermediate intensity. The proportions of the ThB+ B cells and thymocytes do not differ significantly among strains.
Host Species: Rat
Clone: 49-H4
Isotype: IgG2c
Cross Reactivity: Mouse
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Specifications
Specifications
| Antigen | Ly-6D |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 49-H4 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Description | Lymphocyte antigen 6 complex, locus D; Thb; Thymocyte B-cell antigen; Ly-61 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Rat |
| Immunogen | BALB/c mouse plasmacytoma MOPC-104E |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.