Learn More
Ly-6G and Ly-6C Rat anti-Mouse, FITC, Clone: RB6-8C5, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 553127
Description
The RB6-8C5 antibody reacts with a common epitope on Ly-6G and Ly-6C, previously known as the myeloid differentiation antigen Gr-1. In the bone marrow, the level of antigen expression is directly correlated with granulocyte differentiation and maturation.
Host Species: Rat
Clone: RB6-8C5
Isotype: IgG2b, κ
Species Reactivity: Mouse
Flow Cytometry
Specifications
| Ly-6G and Ly-6C | |
| Monoclonal | |
| 0.5mg/mL | |
| Ly6c, Lymphocyte antigen 6C2; Lymphocyte antigen 6G, Ly6g, Gr-1 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| RB6-8C5 | |
| FITC | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Not Reported | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.