missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M13 Phage coat protein Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA126758
Additional Details : Weight : 0.01000kg
Description
PA1-26758 specifically detects phage coat proteins of M13 or fd phage. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
M13 Phage coat protein Polyclonal antibody specifically detects M13 Phage coat protein in Virus samples. It is validated for ELISA, Immunohistochemistry.Specifications
M13 Phage coat protein | |
Polyclonal | |
Unconjugated | |
Rabbit | |
IgG fraction | |
RUO | |
Virus | |
Antibody | |
IgG |
ELISA, Immunohistochemistry | |
6.9 mg/mL | |
0.01M PBS with 15mM sodium azide; pH 7.4 | |
fd bacteriophage. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |