Learn More
MAD2 mitotic arrest deficient-like 2 (yeast), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human MAD2L2 protein.
Brand: Abnova H00010459-D01P.100ug
Description
The protein encoded by this gene is a component of the mitotic spindle assembly checkpoint that prevents the onset of anaphase until all chromosomes are properly aligned at the metaphase plate. The encoded protein, which is similar to MAD2L1, is capable of interacting with ADAM9, ADAM15, REV1, and REV3 proteins. [provided by RefSeq
Sequence: MTTLTRQDLNFGQVVADVLCEFLEVAVHLILYVREVYPVGIFQKRKKYNVPVQMSCHPELNQYIQDTLHCVKPLLEKNDVEKVVVVILDKEHRPVEKFVFEITQPPLLSISSDSLLSHVEQLLRAFILKISVCDAVLDHNPPGCTFTVLVHTREAATRNMEKIQVIKDFPWILADEQDVHMHDPRLIPLKTMTSDILKMQLYVEERAHKGSSpecifications
| MAD2 mitotic arrest deficient-like 2 (yeast) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human MAD2L2 protein. | |
| MAD2L2 | |
| MAD2B/REV7 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC015244 | |
| MAD2L2 | |
| MAD2L2 (AAH15244.1, 1 a.a. ∼ 211 a.a) full-length human protein. | |
| 100 μg | |
| Cell Cycle | |
| 10459 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.