Learn More
microtubule-associated protein, RP/EB family, member 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MAPRE1 protein.
Brand: Abnova H00022919-B01.50uL
Description
The protein encoded by this gene was first identified by its binding to the APC protein which is often mutated in familial and sporadic forms of colorectal cancer. This protein localizes to microtubules, especially the growing ends, in interphase cells. During mitosis, the protein is associated with the centrosomes and spindle microtubules. The protein also associates with components of the dynactin complex and the intermediate chain of cytoplasmic dynein. Because of these associations, it is thought that this protein is involved in the regulation of microtubule structures and chromosome stability. This gene is a member of the RP/EB family. [provided by RefSeq
Sequence: MAVNVYSTSVTSDNLSRHDMLAWINESLQLNLTKIEQLCSGAAYCQFMDMLFPGSIALKKVKFQAKLEHEYIQNFKILQAGFKRMGVDKIIPVDKLVKGKFQDNFEFVQWFKKFFDANYDGKDYDPVAARQGQETAVAPSLVAPALNKPKKPLTSSSAAPQRPISTQRTAAAPKAGPGVVRKNPGVGNGDDEAAELMQQVNVLKLTVEDLEKERDFYFGKLRNIELICQENEGENDPVLQRIVDILYATDEGFVIPDEGGPQEEQEEYSpecifications
| microtubule-associated protein, RP/EB family, member 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human MAPRE1 protein. | |
| MAPRE1 | |
| EB1/MGC117374/MGC129946 | |
| Mouse | |
| 50 μL | |
| Cell Cycle | |
| Primary | |
| Human, Mouse, Rat | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_012325 | |
| MAPRE1 | |
| MAPRE1 (NP_036457, 1 a.a. ∼ 268 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 22919 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.