missing translation for 'onlineSavingsMsg'
Learn More

Mast Cell Chymase Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA528317

Product Code. 13462427

  • € 568.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PA5-28317 targets Mast Cell Chymase in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-28317 immunogen is recombit fragment corresponding to a region within amino acids 1 and 218 of Human Mast Cell Chymase. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene product is a chymotryptic serine proteinase that belongs to the peptidase family S1. It is expressed in mast cells and thought to function in the degradation of the extracellular matrix, the regulation of submucosal gland secretion, and the generation of vasoactive peptides. In the heart and blood vessels, this protein, rather than angiotensin converting enzyme, is largely responsible for converting angiotensin I to the vasoactive peptide angiotensin II. Angiotensin II has been implicated in blood pressure control and in the pathogenesis of hypertension, cardiac hypertrophy, and heart failure. Thus, this gene product is a target for cardiovascular disease therapies. This gene maps to 14q11.2 in a cluster of genes encoding other proteases.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mast Cell Chymase
Polyclonal
Unconjugated
P23946
CMA1
Recombit fragment corresponding to a region within amino acids 1 and 218 of Human Mast Cell Chymase
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
CMA1
CYH, MCT1, chymase, alpha-chymase, chymase, chymase 1 preproprotein transcript E, chymase 1 preproprotein transcript I, chymase, heart, chymase, mast cell, mast cell protease I
Rabbit
Antigen affinity chromatography
RUO
1215
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.