Learn More
myeloid cell leukemia sequence 1 (BCL2-related), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MCL1.
Brand: Abnova H00004170-A02.50uL
Description
The protein encoded by this gene belongs to the Bcl-2 family. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified. The longer gene product (isoform 1) enhances cell survival by inhibiting apoptosis while the alternatively spliced shorter gene product (isoform 2) promotes apoptosis and is death-inducing. [provided by RefSeq
Sequence: GNNTSTDGSLPSTPPPAEEEEDELYRQSLEIISRYLREQATGAKDTKPMGRSGATSRKALETLRRVGDGVQRNHETAFQGMLRKLDIKNEDDVKSLSRVMSpecifications
| myeloid cell leukemia sequence 1 (BCL2-related) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant MCL1. | |
| MCL1 | |
| BCL2L3/EAT/MCL1L/MCL1S/MGC104264/MGC1839/TM | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_021960 | |
| MCL1 | |
| MCL1 (NP_068779, 151 a.a. ∼ 250 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 4170 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.