missing translation for 'onlineSavingsMsg'
Learn More

MICAL1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Product Code. 16103289
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16103289 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16103289 Supplier Abnova Supplier No. H00064780B01P.50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a full-length human MICAL1 protein.

Sequence: MASPTSTNPAHAHFESFLQAQLCQDVLSSFQELCGALGLEPGGGLPQYHKIKDQLNYWSAKSLWTKLDKRAGQPVYQQGRACTSTKCLVVGAGPCGLRVAVELALLGARVVLVEKRTKFSRHNVLHLWPFTIHDLRALGAKKFYGRFCTGTLDHISIRQLQLLLLKVALLLGVEIHWGVTFTGLQPPPRKGSGWRAQLQPNPPAQLANYEFDVLISAAGGKFVPEGFKVREMRGKLAIGITANFVNGRTVEETQVPEISGVARIYNQSFFQSLLKATGIDLENIVYYKDDTHYFVMTAKKQCLLRLGVLRQPFWPLGTGVARGFLAAFDAAWMVKRWAEGAESLEVLAERESLYQLLSQTSPENMHRNVAQYGLDPATRYPNLNLRAVTPNQVRDLYDVLAKEPVQRNNDKTDTGMPATGSAGTQEELLRWCQEQTAGYPGVHVSDLSSSWADGLALCALVYRLQPGLLEPSELQGLGALEATAWALKVAENELGITPVVSAQAVVAGSDPLGLIAYLSHFHSAFKSMAHSPGPVSQTSPGTSSAVLFLSKLQRTLQRSRAKENAEDAGGKKLRLEMEAETPSTEVPPDPEPGVPLTPPSQHQEAGAGDLCALCGEHLYVLERLCVNGHFFHRSCFRCHTCEATLWPGGYEQHPGDGHFYCLQHLPQTDHKKEGSDRGPESPELPTPSENSMPPGLSTPTASQEGAGPVPDPSQPTRRQIRLSSPERQRLSSLNLTPDPEMEPPPKPPRSCSALARHALESSFVGWGLPVQSPQALVAMEKEEKESPFSSEEEEEDVPLDSDVEQALQTFAKTSGTMNNYPTWRRTLLRRAKEEEMKRFCKAQTIQRRLNEIEAALRELEAEGVKLELALRRQSSSPEQQKKLWVGQLLQLVDKKNSLVAEEAELMITVQELNLEEKQWQLDQELRGYMNREENLKTAADRQAEDQVLRKLVDLVNQRDALIRFQEERRLSELALGTGAQG

Specifications

Antigen MICAL1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human MICAL1 protein.
Formulation PBS with no preservative; pH 7.4
Gene MICAL1
Gene Accession No. BC009972.2
Gene Alias DKFZp434B1517/FLJ11937/FLJ21739/MICAL/MICAL-1/NICAL
Gene Symbols MICAL1
Host Species Mouse
Immunogen MICAL1 (AAH09972.2, 1 a.a. ∼ 981 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64780
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.