missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP kinase interacting serine/threonine kinase 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MKNK2.
Brand: Abnova H00002872-A02.50uL
This item is not returnable.
View return policy
Description
Sequence: GRCGSDCGWDRGEACPACQNMLFESIQEGKYEFPDKDWAHISCAAKDLISKLLVRDAKQRLSAAQVLQHPWVQGCAPENTLPTPMVLQRWDSHFLLPPHPCRIHVRPGGLVRTVTVNESpecifications
| MAP kinase interacting serine/threonine kinase 2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant MKNK2. | |
| MKNK2 | |
| GPRK7/MNK2 | |
| Mouse | |
| 50 μL | |
| Kinases | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC018345 | |
| MKNK2 | |
| MKNK2 (AAH18345, 41 a.a. ∼ 158 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 2872 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction