missing translation for 'onlineSavingsMsg'
Learn More

myeloid/lymphoid or mixed-lineage leukemia 4, Mouse, Clone: 1F6, Abnova™

Product Code. 16151256
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16151256 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16151256 Supplier Abnova Supplier No. H00009757M01.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against a partial recombinant MLL4.

This gene encodes a protein which contains multiple domains including a CXXC zinc finger, three PHD zinc fingers, two FY-rich domains, and a SET (suppressor of variegation, enhancer of zeste, and trithorax) domain. The SET domain is a conserved C-terminal domain that characterizes proteins of the MLL (mixed-lineage leukemia) family. This gene is ubiquitously expressed in adult tissues. It is also amplified in solid tumor cell lines, and may be involved in human cancer. Two alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene, however, the full length nature of the shorter transcript is not known. [provided by RefSeq

Sequence: KVAASMPLSPGGQMEEVAGAVKQISDRGPVRSEDESVEAKRERPSGPESPVQGPRIKHVCRHAAVALGQARAMVPEDVPRLSALPLRDRQDL

Specifications

Antigen myeloid/lymphoid or mixed-lineage leukemia 4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant MLL4.
Formulation PBS with no preservative; pH 7.4
Gene MLL4
Gene Accession No. NM_014727
Gene Alias HRX2/KIAA0304/MLL2/TRX2/WBP7
Gene Symbols MLL4
Host Species Mouse
Immunogen MLL4 (NP_055542, 813 a.a. ∼ 904 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 9757
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.