Learn More
matrix metallopeptidase 28, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human MMP28 protein.
Brand: Abnova H00079148-D01P.100ug
Description
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix for both normal physiological processes, such as embryonic development, reproduction and tissue remodeling, and disease processes, such as asthma and metastasis. This gene encodes a secreted enzyme that degrades casein. Its expression pattern suggests that it plays a role in tissue homeostasis and in wound repair. Transcript variants encoding different isoforms have been described. [provided by RefSeq
Sequence: MVARVGLLLRALQLLLWGHLDAQPAERGGQELRKEAEAFLEKYGYLNEQVPKAPTSTRFSDAIRAFQWVSQLPVSGVLDRATLRQMTRPRCGVTDTNSYAAWAERISDLFARHRTKMRRKKRFAKQGNKWYKQHLSYRLVNWPEHLPEPAVRGAVRAAFQLWSNVSALEFWEAPATGPADIRLTFFQGDHNDGLGNAFDGPGGALAHAFLPRRGEAHFDQDERWSLSRRRGRNLFVVLAHEIGHTLGLTHSPAPRALMAPYYKRLGRDALLSWDDVLAVQSLYGKPLGGSVAVQLPGKLFTDFETWDSYSPQGRRPETQGPKYCHSSFDAITVDRQQQLYIFKGSHFWEVAADGNVSEPRPLQERWVGLPPNIEAAAVSLNDGDFYFFKVQSVSpecifications
| matrix metallopeptidase 28 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human MMP28 protein. | |
| MMP28 | |
| EPILYSIN/MM28/MMP-28/MMP25 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 79148 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC002631.1 | |
| MMP28 | |
| MMP28 (AAH02631, 1 a.a. ∼ 393 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.