missing translation for 'onlineSavingsMsg'
Learn More

Anti-Mouse B7-H3/CD276 (MJ18), Proteintech

Product Code. 30237515 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30237515 500 μg 500µg
30233334 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30237515 Supplier Proteintech Supplier No. 652211IG500UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

The B7-H3/CD276 antibody from Proteintech is a rat monoclonal antibody generated with mouse igg2a fc-cd276 (aa 1-242) fusion protein of mouse B7-H3/CD276. This antibody recognizes mouse antigen. The B7-H3/CD276 antibody has been validated for the following applications: FC analysis.

Specifications

Antigen B7-H3/CD276
Applications Flow Cytometry
Classification Monoclonal
Clone MJ18
Concentration 0.5 mg/mL
Conjugate Unconjugated
Host Species Rat
Immunogen Mouse IgG2a Fc-CD276 (aa 1-242) fusion protein
Purification Method Protein G purification
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102657
Target Species Mouse
Content And Storage Store at 2-8°C. Stable for one year after shipment.
Product Type Flow Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.