missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Mouse CD134/OX40 (OX-86), Proteintech
Rat Monoclonal Antibody
Brand: Proteintech 65136-1-IG-100UG
This item is not returnable.
View return policy
Description
The CD134/OX40 antibody from Proteintech is a rat monoclonal antibody generated with n/a of mouse CD134/OX40. This antibody recognizes mouse antigen. The CD134/OX40 antibody has been validated for the following applications: FC analysis.Specifications
| CD134/OX40 | |
| Monoclonal | |
| 500 μg/mL | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Flow Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| OX-86 | |
| Unconjugated | |
| Affinity purification | |
| RUO | |
| 22163 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction