missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Mouse CD41 (MWReg30), Proteintech
Rat Monoclonal Antibody
€ 60.00 - € 151.00
Specifications
| Antigen | CD41 |
|---|---|
| Clone | MWReg30 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234373
|
Proteintech
65189-1-IG-100UG |
100 μg |
€ 60.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234142
|
Proteintech
65189-1-IG-500UG |
500 μg |
€ 151.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD41 antibody from Proteintech is a rat monoclonal antibody generated with mouse platelets of mouse CD41. This antibody recognizes mouse antigen. The CD41 antibody has been validated for the following applications: FC analysis.Specifications
| CD41 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 16399 | |
| IgG1 κ | |
| Purified by protein-G affinity chromatography | |
| Flow Antibody |
| MWReg30 | |
| Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Mouse platelets | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title