missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD45, Clone: PD7/26/16 and 2B11, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44176C
This item is not returnable.
View return policy
Description
GSTO1 Monoclonal specifically detects GSTO1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| CD45 | |
| Monoclonal | |
| Unconjugated | |
| BALB/C mice were immunized with human peripheral blood | |
| RUO | |
| 5788 | |
| Human | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| PD7/26/16 and 2B11 | |
| Mouse | |
| 6 mL | |
| Primary | |
| The human leukocyte common antigen is a family of five or more high molecular weight glycoproteins of MW 180, 190, 205 and 220 kD, present on the surface of the majority of human leucocytes. This labels lymphoid cells. This antibody labels lymphoid cells. It labels neoplastic B and T cells in non-Hodgkin's lymphoma and in leukemias of B and T cell types. Cross-reacts with Human. Not yet tested in other species. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction