Learn More
Anti-Mouse CD81 (EAT2), Proteintech
Hamster Monoclonal Antibody
Brand: Proteintech 65220-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD81 antibody from Proteintech is a armenian hamster monoclonal antibody generated with cd81+ mouse b lymphoma 38c13 of mouse CD81. This antibody recognizes mouse antigen. The CD81 antibody has been validated for the following applications: FC analysis.Specifications
| CD81 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Armenian Hamster | |
| Protein G purification | |
| RUO | |
| 12520 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| EAT2 | |
| Unconjugated | |
| CD81+ mouse B lymphoma 38C13 | |
| 500 μg | |
| Primary | |
| Mouse | |
| Flow Antibody | |
| IgG |