missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse Lambda Light Chain Secondary Antibody, AP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA186022
Additional Details : Weight : 0.01000kg
Description
Mouse Lambda Light Chain Polyclonal antibody specifically detects Mouse Lambda Light Chain in Mouse samples. It is validated for ELISA.Specifications
Mouse Lambda Light Chain | |
Polyclonal | |
TBS with 1mM MgCl2, 50% glycerol and <0.1% sodium azide | |
Goat | |
Affinity chromatography | |
RUO | |
Mouse | |
Liquid |
ELISA | |
Alkaline Phosphatase (AP) | |
Iglc | |
Murine paraproteins with lambda 1 and lambda 2 light chains. | |
1 mL | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |