Learn More
Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, DSB-X Biotin, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen D20691
Additional Details : Weight : 0.00450kg
Description
DSB-X biotin is a derivative of desthiobiotin, a stable biotin percursuor. DSB-X biotin utilizes a seven-atom spacer to increase the available of the DSB-X biotin conjugate to bind in the deep biotin-binding pocket of streptpavidin or avidin. Whereas harsh chaotropic agents and low pH are required to dissociate a biotin complex from avidin or steptavidin, binding can be reversed by adding excess D-desthiobiotin or natural D-biotin at neutral pH and at room temperature.
Mouse IgG (H+L) Highly Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG (H+L) Highly Cross-Adsorbed in Mouse samples. It is validated for Immunocytochemistry.Specifications
Mouse IgG (H+L) Highly Cross-Adsorbed | |
Polyclonal | |
DSB-X Biotin | |
Goat | |
purified | |
RUO | |
Mouse | |
Liquid |
Immunocytochemistry | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
4° C, store in dark | |
IgG |