Learn More
Goat anti-Mouse IgG, IgM, IgA (H+L) Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA184388
Additional Details : Weight : 0.01000kg
Description
Goat anti Mouse IgG/A/M (Human Adsorbed) antibody recognizes all subclasses of murine immunoglobulin and has been adsorbed against human, bovine and hamster immunoglobulins.
Mouse IgG, IgM, IgA (H+L) Polyclonal antibody specifically detects Mouse IgG, IgM, IgA (H+L) in Mouse samples. It is validated for ELISA, Western Blot.Specifications
Mouse IgG, IgM, IgA (H+L) | |
Polyclonal | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Mouse | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.01% thimerosal | |
Purified murine immunoglobulins. | |
1 mg | |
Secondary | |
4° C, do not freeze | |
IgG |