Learn More
Anti-Mouse Ly-6G (1A8), Proteintech
Rat Monoclonal Antibody
Brand:  Proteintech 65078-1-IG-100UG
 View more versions of this product
Product Code. 30236559
  • € 38.00 / 100µg
Description
The Ly-6G antibody from Proteintech is a rat monoclonal antibody generated with ly-6g transfected el-4j cell line of mouse Ly-6G. This antibody recognizes mouse antigen. The Ly-6G antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
Ly-6G
Monoclonal
500 μg/mL
Rat
Affinity purification
RUO
546644
Store at 2-8°C. Stable for one year after shipment.
Liquid
Flow Cytometry
1A8
Unconjugated
Ly-6G transfected EL-4J cell line
100 μg
Primary
Mouse
Flow Antibody
IgG2a κ