missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Mouse NK1.1 (CD161) (PK136), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65138-1-IG-100UG
This item is not returnable.
View return policy
Description
The NK1.1 (CD161) antibody from Proteintech is a mouse monoclonal antibody generated with nk-1+ cells from mouse spleen and bone marrow of mouse NK1.1 (CD161). This antibody recognizes mouse antigen. The NK1.1 (CD161) antibody has been validated for the following applications: FC analysis.Specifications
| NK1.1 (CD161) | |
| Monoclonal | |
| 0.5 mg/mL | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 17057 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| PK136 | |
| Unconjugated | |
| NK-1+ cells from mouse spleen and bone marrow | |
| 100 μg | |
| Primary | |
| Mouse | |
| Flow Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction