missing translation for 'onlineSavingsMsg'
Learn More

Anti-Mouse PD-1/CD279 (RMP1-30), Proteintech

Rat Monoclonal Antibody

€ 71.53 - € 214.60

Specifications

Antigen PD-1/CD279
Clone RMP1-30
Concentration 0.5 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30233967
View Documents
Proteintech
65142-1-IG-100UG
100 μg
€ 71.53
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30233162
View Documents
Proteintech
65142-1-IG-500UG
500 μg
€ 214.60
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The PD-1/CD279 antibody from Proteintech is a rat monoclonal antibody generated with mouse pd-1 transfected bhk cells of mouse PD-1/CD279. This antibody recognizes mouse antigen. The PD-1/CD279 antibody has been validated for the following applications: FC analysis.
Specifications

Specifications

PD-1/CD279
0.5 mg/mL
Monoclonal
Liquid
RUO
18566
IgG2b κ
Protein A purification
Flow Antibody
RMP1-30
Flow Cytometry
Unconjugated
Rat
Mouse
Mouse PD-1 transfected BHK cells
Primary
Store at 2-8°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.