Learn More
myosin IIIA, Mouse, Clone: 8H2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant MYO3A.
Brand: Abnova H00053904-M08.50ug
Description
The protein encoded by this gene belongs to the myosin superfamily. Myosins are actin-dependent motor proteins and are categorized into conventional myosins (class II) and unconventional myosins (classes I and III through XV) based on their variable C-terminal cargo-binding domains. Class III myosins, such as this one, have a kinase domain N-terminal to the conserved N-terminal motor domains and are expressed in photoreceptors. The protein encoded by this gene plays an important role in hearing in humans. Three different recessive, loss of function mutations in the encoded protein have been shown to cause nonsyndromic progressive hearing loss. Expression of this gene is highly restricted, with the strongest expression in retina and cochlea. [provided by RefSeq
Sequence: HEEINNIKKKDNKDSKATSEREACGLAIFSKQISKLSEEYFILQKKLNEMILSQQLKSLYLGVSHHKPINRRVSSQQCLSGVCKGEEPKILSpecifications
| myosin IIIA | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_017433 | |
| MYO3A | |
| MYO3A (NP_059129, 1400 a.a. ∼ 1490 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 50 μg | |
| Kinases | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 8H2 | |
| Mouse monoclonal antibody raised against a partial recombinant MYO3A. | |
| MYO3A | |
| DFNB30 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 53904 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.