Learn More
N-myc downstream regulated 1, Mouse, Clone: 2D7, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant NDRG1.
Brand: Abnova H00010397-M03.100ug
Description
This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein involved in stress responses, hormone responses, cell growth, and differentiation. It is necessary for p53-mediated caspase activation and apoptosis. Mutation in this gene has been reported to be causative for hereditary motor and sensory neuropathy-Lom. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq
Sequence: MSREMQDVDLAEVKPLVEKGETITGLLQEFDVQEQDIETLHGSVHVTLCGTPKGNRPVILTYHDIGMNHKTCYNPLFNYEDMQEITQHFAVCHVDAPGQQDGAASFPAGYMYPSMDQLAEMLPGVLQQFGLKSIIGMGTGAGAYILTRFALNNPEMVEGLVLINVNPCAEGWMDWAASKISGWTQALPDMVVSHLFGKEEMQSNVEVVHTYRQHIVNDMNPGNLHLFINAYNSRRDLEIERPMPGTHTVTLQCPALLVVGDSSPAVDAVVECNSKLDPTKTTLLKMADCGGLPQISQPAKLAEAFKYFVQGMGYMPSASMTRLMRSRTASGSSVTSLDGTRSRSHTSEGTRSRSHTSEGTRSRSHTSEGAHLDITPNSGAAGNSAGPKSMEVSCSpecifications
| N-myc downstream regulated 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC003175 | |
| NDRG1 | |
| NDRG1 (AAH03175, 1 a.a. ∼ 394 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 2D7 | |
| Mouse monoclonal antibody raised against a full length recombinant NDRG1. | |
| NDRG1 | |
| CAP43/CMT4D/DRG1/GC4/HMSNL/NDR1/NMSL/PROXY1/RIT42/RTP/TARG1/TDD5 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 10397 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.