missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFV1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant NDUFV1.
Brand: Abnova H00004723-A01.50uL
This item is not returnable.
View return policy
Description
The NDUFV1 gene encodes the 51kDa subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.[supplied by OMIM
Sequence: KAIARLIEFYKHESCGQCTPCREGVDWMNKVMARFVRGDARPAEIDSLWEISKQIEGHTICALGDGAAWPVQGLIRHFRPELEERMQRFAQQHQARQAASSpecifications
| NDUFV1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant NDUFV1. | |
| NDUFV1 | |
| CI-51kD/UQOR1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_007103 | |
| NDUFV1 | |
| NDUFV1 (NP_002466, 365 a.a. ∼ 464 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 4723 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction