Learn More
NDUFV3, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human NDUFV3 protein.
Brand: Abnova H00004731-D01P.100ug
Description
The protein encoded by this gene is one of at least forty-one subunits that make up the NADH-ubiquinone oxidoreductase complex. This complex is part of the mitochondrial respiratory chain and serves to catalyze the rotenone-sensitive oxidation of NADH and the reduction of ubiquinone. The encoded protein is one of three proteins found in the flavoprotein fraction of the complex. The specific function of the encoded protein is unknown. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MAAPCLLRQGRAGALKTMLQEAQVFRGLASTVSLSAESGKSEKGQPQNSKKQSPPKKPAPVPAEPFDNTTYKNLQHHDYSTYTFLDLNLELSKFRMPQPSSGRESPRHSpecifications
| NDUFV3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human NDUFV3 protein. | |
| NDUFV3 | |
| CI-9KD | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4731 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001001503.1 | |
| NDUFV3 | |
| NDUFV3 (NP_001001503.1, 1 a.a. ∼ 108 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.