missing translation for 'onlineSavingsMsg'
Learn More

NIMA (never in mitosis gene a)- related kinase 11, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Product Code. 16163859
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16163859 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16163859 Supplier Abnova Supplier No. H00079858D01P.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against a full-length human NEK11 protein.

This gene encodes a member of the never in mitosis gene A family of kinases. The encoded protein localizes to the nucleoli, and may function with NEK2A in the S-phase checkpoint. The encoded protein appears to play roles in DNA replication and response to genotoxic stress. Alternatively spliced transcript variants have been described

Sequence: MLKFQEAAKCVSGSTAISTYPKTLIARRYVLQQKLGSGSFGTVYLVSDKKAKRGEELKVLKEISVGELNPNETVQANLEAQLLSKLDHPAIVKFHASFVEQDNFCIITEYCEGRDLDDKIQEYKQAGKIFPENQIIEWFIQLLLGVDYMHERRILHRDLKSKNVFLKNNLLKIGDFGVSRLLMGSCDLATTLTGTPHYMSPEALKHQGYDTKSDIWSLACILYEMCCMNHAFAGSNFLSIVLKIVEGDTPSLPERYPKELNAIMESMLNKNPSLRPSAIEILKIPYLDEQLQNLMCRYSEMTLEDKNLDCQKEAAHIINAMQKRIHLQTLRALSEVQKMTPRERMRLRKLQAADEKARKLKKIVEEKYEENSKRMQELRSRNFQQLSVDVLHEKTHLKGMEEKEEQPEGRLSCSPQDEDEERWQGREEESDEPTLENLPESQPIPSMDLHELESIVEDATSDLGYHGDCNLISLDEYWKNDK

Specifications

Antigen NIMA (never in mitosis gene a)- related kinase 11
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human NEK11 protein.
Formulation PBS with no preservative; pH 7.4
Gene NEK11
Gene Accession No. BC028587.2
Gene Alias FLJ23495
Gene Symbols NEK11
Host Species Rabbit
Immunogen NEK11 (AAH28587.1, 1 a.a. ∼ 482 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Kinases
Primary or Secondary Primary
Gene ID (Entrez) 79858
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.