Learn More
sialidase 2 (cytosolic sialidase), Mouse, Clone: 2E5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant NEU2.
Brand: Abnova H00004759-M04.100ug
Description
This gene belongs to a family of glycohydrolytic enzymes which remove sialic acid residues from glycoproteins and glycolipids. Expression studies in COS7 cells confirmed that this gene encodes a functional sialidase. Its cytosolic localization was demonstrated by cell fractionation experiments. [provided by RefSeq
Sequence: AYRKLHPIQRPIPSAFCFLSHDHGRTWARGHFVAQDTLECQVAEVETGEQRVVTLNARSHLRARVQAQSTNDGLDFQESQLVKKLVEPPSpecifications
| sialidase 2 (cytosolic sialidase) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005383 | |
| NEU2 | |
| NEU2 (NP_005374, 180 a.a. ∼ 268 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 4759 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a λ |
| ELISA, Western Blot | |
| 2E5 | |
| Mouse monoclonal antibody raised against a partial recombinant NEU2. | |
| NEU2 | |
| MGC129579/SIAL2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.