Learn More
nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant NFATC1.
Brand: Abnova H00004772-A01.50uL
Description
The product of this gene is a component of the nuclear factor of activated T cells DNA-binding transcription complex. This complex consists of at least two components: a preexisting cytosolic component that translocates to the nucleus upon T cell receptor (TCR) stimulation, and an inducible nuclear component. Proteins belonging to this family of transcription factors play a central role in inducible gene transcription during immune response. The product of this gene is an inducible nuclear component. It functions as a major molecular target for the immunosuppressive drugs such as cyclosporin A. Five transcript variants encoding distinct isoforms have been identified for this gene. Different isoforms of this protein may regulate inducible expression of different cytokine genes. [provided by RefSeq
Sequence: IKTEPTDDYEPAPTCGPVSQGLSPLPRPYYSQQLAMPPDPSSCLVAGFPPCPQRSTLMPAAPGVSPKLHDLSPAAYTKGVASPGHCHLGLPQPAGEAPAVQDVPRPVATHSpecifications
| nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant NFATC1. | |
| NFATC1 | |
| MGC138448/NF-ATC/NFAT2/NFATc | |
| Mouse | |
| 50 μL | |
| Transcription Regulation | |
| 4772 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_006162 | |
| NFATC1 | |
| NFATC1 (NP_006153, 701 a.a. ∼ 810 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.