Learn More
neuroglobin, Mouse, Clone: 1A11-A9, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant NGB.
Brand: Abnova H00058157-M02.100ug
Description
This gene encodes an oxygen-binding protein that is distantly related to members of the globin gene family. It is highly conserved among other vertebrates. It is expressed in the central and peripheral nervous system where it may be involved in increasing oxygen availability and providing protection under hypoxic/ischemic conditions. [provided by RefSeq
Sequence: MERPEPELIRQSWRAVSRSPLGHGTVLFARLFALEPDLLPLFQYNCRQFSSPEDCLSSPEFLDHIRKVMLVIDAAVTNVEDLSSLEEYLASLGRKHRAVGVKLSSFSTVGESLLYMLEKCLGPAFTPATRAAWSQLYGAVVQAMSRGWDGESpecifications
| neuroglobin | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC032509 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1A11-A9 | |
| Mouse monoclonal antibody raised against a full length recombinant NGB. | |
| NGB | |
| NGB | |
| NGB (AAH32509, 1 a.a. ∼ 151 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 58157 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.