missing translation for 'onlineSavingsMsg'
Learn More

NMDAR2A (N-Methyl D-Aspartate) Mouse, Unlabeled, Clone: 5, BD

Product Code. 15896149
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15896149 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15896149 Supplier BD Biosciences Supplier No. 612286

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The majority of synapses in the central nervous system utilize glutamate as a neurotransmitter to produce rapid neuronal excitation. Glutamate has a diverse array of receptors that can be categorized into two groups, ionotropic and metabotropic. The ionotropic receptors are subdivided into two distinct types: 1) receptors for N-methyl D-aspartate (NMDAR) and 2) non-NMDA receptors for AMPA and kainate. Four types of NMDAR2 have been identified: NMDAR2A (NR2A), NR2B, NR2C, and NR2D. NR2A and NR2B contain a C-terminal extension (> 600 amino acids) that has small, scattered regions of conserved sequence. In embryonic stages, NR2B is expressed throughout the brain, while NR2D is found in diencephalon and brainstem. After birth, NR2A is found throughout the brain, and NR2C is expressed in cerebellum. At this time, NR2B expression becomes restricted to the forebrain, and NR2D expression is strongly reduced. Deletion of the most widespread postnatal NR subunit, NR2A, does not have adverse affects on brain development, but reduces long-term potentiation (LTP) in the hippocampus and impairs spatial and contextual learning. Thus, NR2A may be critical for postnatal learning.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR2A (N-Methyl D-Aspartate)
Applications Western Blot
Classification Monoclonal
Clone 5
Concentration 250μg/mL
Conjugate Unconjugated
Description N-Methyl D-Aspartate
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse NMDAR2A aa. 1093-1214
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.