missing translation for 'onlineSavingsMsg'
Learn More

Nociceptin Rabbit anti-Non-human primate, Rat, Polyclonal, Invitrogen™

Product Code. 11541313
Encompass
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11541313 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11541313 Supplier Invitrogen Supplier No. PA127972

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-27972 detects Nociceptin from rat, non-human primate samples. PA1-27972 has been successfully used in immunohistochemistry (frozen tissue) applications. The PA1-27972 immunogen is: Synthetic peptide: FGGFTGARKSARKLANQ, corresponding to amino acids 122-138 of Rat Orphanin FQ. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Nociceptin is the ligand of the opioid receptor-like receptor (OPRL1). It may act as a transmitter in the brain by modulating nociceptive and locomotor behavior. May be involved in neuronal differentiation and development.

Specifications

Antigen Nociceptin
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Gene Pnoc
Gene Accession No. Q62923
Gene Alias Orphanin FQ, PPNOC
Gene Symbols PNOC
Host Species Rabbit
Immunogen synthetic peptide FGGFTGARKSARKLANQ, corresponding to amino acids 122-138 of Rat Orphanin FQ.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25516
Target Species Non-human Primate, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.