missing translation for 'onlineSavingsMsg'
Learn More

Occludin Mouse, Unlabeled, Clone: 19, BD

Product Code. 15865789
Change view
Click to view available options
Quantity:
150 μg
50 μg
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15865789 50 μg 1 set
15875789 150 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15865789 Supplier BD Biosciences Supplier No. 611090

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tight junctions (zonulae occludens) are critical to the maintenance of cell polarity and intercellular barriers between epithelial and endothelial cells. Among the barriers formed by the tight junctions is the critical blood-brain barrier. Protein components of the tight junctions include actin filaments, symplekin, occludin, Rab3B, AF-6, 7H6, ZO-1 and ZO-2. The human occludin protein is 522 amino acids with four transmembrane domains in the first half of the protein. Two glycine/tyrosine-rich extracellular loops are involved in cell-cell interactions. In the cytoplasm, occludin interacts with ZO-1 through its intracellular C-terminus. A possible function of ZO-1 may be to link the adherens components, like occludin, to the submembraneous actin cytoskeleton. Experimental breakdown of the tight junctions at the blood-brain barrier results in redistribution of vinculin and loss of ZO-1 and Occludin. Taken together, these data indicate that occludin and ZO-1 are critical elements in the formation of tight junctions and may also serve an important role in signaling and tumor suppression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Occludin
Applications Western Blot
Classification Monoclonal
Clone 19
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse Occludin aa. 396-507
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.