Learn More
origin recognition complex, subunit 4-like (yeast), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human ORC4L protein.
Brand: Abnova H00005000-D01P.100ug
Description
The origin recognition complex (ORC) is a highly conserved six subunit protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is a subunit of the ORC complex. It has been shown to form a core complex with ORC2L, -3L, and -5L. Three alternatively spliced transcript variants encoding the same protein have been reported. [provided by RefSeq
Sequence: MSSRKSKSNSLIHTECLSQVQRILRERFCRQSPHSNLFGVQVQYKHLSELLKRTALHGESNSVLIIGPRGSGKTMLINHALKELMEIEEVSENVLQVHLNGLLQINDKIALKEITRQLNLENVVGDKVFGSFAENLSFLLEALKKGDRTSSCPVIFILDEFDLFAHHKNQTLLYNLFDISQSAQTPIAVIGLTCRLDILELLEKRVKSRFSHRQIHLMNSFGFPQYVKIFKEQLSLPAEFPDKVFAEKWNENVQYLSEDRSVQEVLQKHFNISKNLRSLHMLLMLALNRVTASHPFMTAVDLMEASQLCSMDSKANIVHGLSVLEICLIIAMKHLNDIYEEEPFNFQMVYNEFQKFVQRKAHSVYNFEKPVVMKAFEHLQQLELIKPMERTSGNSQREYQLMKLLLDNTQIMNALQKYPNCPTDVRQWATSSLSWLSpecifications
| origin recognition complex, subunit 4-like (yeast) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human ORC4L protein. | |
| ORC4L | |
| ORC4/ORC4P | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5000 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002552 | |
| ORC4L | |
| ORC4L (NP_002543.2, 1 a.a. ∼ 436 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.