Learn More
anti-OSM, COL4A6 Protein Protein Interaction Antibody Pair, Abnova™
Product Code. 16171114
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16171114 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16171114 Supplier Abnova Supplier No. DI0105.1Set
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
Includes
OSM rabbit purified polyclonal antibody (20μg); COL4A6 mouse monoclonal antibody (40μug); Reagents are sufficient for at least 30-50 assays using recommended protocols
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the OSM protein, and the other against the COL4A6 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
Antigen OSM
Applications In situ PLA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Gene OSM
Gene Alias MGC20461
Gene Symbols OSM
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5008/1288
Includes Antibody pair set content: 1. OSM rabbit purified polyclonal antibody (20 ug) 2. COL4A6 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Type OSM & COL4A6 Protein Protein Interaction Antibody Pair
Form Purified