Learn More
p70 [S6K] Mouse, Unlabeled, Clone: 16, BD
Description
Increased protein synthesis, an obligatory step in cell proliferation, is regulated by the phosphorylation of a number of translational components, such as ribosomal protein S6. p70 S6 Kinase (p70 [S6K]), a cytosolic kinase that is primarily responsible for S6 phosphorylation, belongs to a family of at least six S6 kinases. p70 [S6K] and p85 [S6K] are two isoforms of the same kinase. Although they are encoded by the same gene, p80 [S6K] contains a 23 amino acid N-terminal extension that serves as a nuclear localization sequence. These kinases are located in a p21 ras-independent signaling pathway and are regulated by Ser/Thr phosphorylation in response to mitogenic stimulation. Activation of p70 [S6K] requires sequential phosphorylations within the autoinhibitory domain and at Thr 389. p70 [S6K] associates with PDK-1 and PKC? to form a possible PI3-K signaling complex. The immunosuppressive agent rapamycin forms an intracellular complex with FKBP12 and FRAP (RAFT1) that inhibits phosphorylation of p70 [S6K] at Thr 389 and, in turn, prevents enzyme activation. Thus, p70 [S6K] is an important regulator of cell proliferation and a potential target of agents that modify the immune and proliferative responses. p70 [S6K] has been reported to be observable migrating within a range of 60-70 kD by western blotting.
Host Species: Mouse
Clone: 16
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat p70 [S6K] aa. 2-121
Formula Weight [Chemical]: 60-70kDa
Immunofluorescence, Western Blotting
Specifications
Specifications
| Antigen | p70 [S6K] |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 16 |
| Concentration | 250μg/mL |
| Conjugate | Unconjugated |
| Description | p70 S6 Kinase |
| Formulation | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Rat p70 [S6K] aa. 2-121 |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.