missing translation for 'onlineSavingsMsg'
Learn More

PAI-1 Mouse, Unlabeled, Clone: 41, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 612025

 View more versions of this product

Product Code. 15856079

  • € 451.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Fibrinolysis is regulated by the plasminogen activators, tissue plasminogen activator (tPA) and urokinase PA (uPA), and by the plasminogen activator-inhibitors (PAIs). Two PAIs include the serpin family members, PAI-1 and PAI-2. PAI-1 is a glycoprotein found in plasma, platelets, endothelial cells, hepatoma cells, and fibrosarcoma cells. Thrombin, endotoxin, and IL-1 induce PAI-1 synthesis in endothelial cells, which is where the major portion of plasma PAI-1 is produced. PAI-2 is a glycoprotein expressed in placenta and monocyte macrophages. The uPA/plasmin system may play a key role in cancer progression through degradation of the extracellular matrix during tumor cell migration. Paradoxically, high levels of PAI-1 are also predictive of poor prognosis of cancer patients. This finding may be a result of the role of plasmin proteolysis in the prevention of tumor vessel assembly. PAI-1 can promote tumor angiogenesis, and the mechanism may involve PAI-induced regulation of plasmin proteolysis during tumor angiogenesis. Thus, PAI-1 is a serpin protease inhibitor that is important for the regulation of plasmin proteolysis during fibrinolysis and extracellular matrix degradation.

Host Species: Mouse
Clone: 41
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human PAI-1 aa. 207-329

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PAI-1
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human PAI-1 aa. 207-329
150 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG1
Western Blot
41
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.