Learn More
protocadherin gamma subfamily A, 5, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human PCDHGA5 protein.
Brand: Abnova H00056110-D01P.100ug
Description
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. [provided by RefSeq
Sequence: MASPPRGWGCGELLLPFMLLGTLCEPGSGQIRYSMPEELDKGSFVGNIAKDLGLEPQELAERGVRIVSRGRTQLFALNPRSGSLVTAGRIDREELCAQSPLCVVNFNILVENKMKIYGVEVEIIDINDNFPRFRDEELKVKVNENAAAGTRLVLPFARDADVGVNSLRSYQLSSNLHFSLDVVSGTDGQKYPELVLEQPLDREKETVHDLLLTALDGGDPVLSGTTHIRVTVLDANDNAPLFTPSEYSVSVPENIPVGTRLLMLTATDPDEGINGKLTYSFRNEEEKISETFQLDSNLGEISTLQSLDYEESRFYLMEVVAQDGGALVASAKVVVTVQDVNDNAPEVILTSLTSSISEDCLPGTVIALFSVHDGDSGENGEIACSIPRNLPFKLEKSVDNYYHLLTTRDLDREETSDYNITLTVMDHGTPPLSTESHIPLKVADVNDNPPNFPQASYSTSVTENNPRGVSIFSVTAHDPDSGDNARVTYSLAEDTFQGAPLSSYVSINSDTGVLYALRSFDYEQLRDLQLWVTASDSGNPPLSSNVSLSLFVLDQNDNTPEILYPALPTDGSTGVELAPRSAEPGYLVTKVVAVDKDSGQNAWLSYRLLKASEPGLFAVGLHTGEVRTARALLDRDALKQSLVVAVEDHGQPPLSATFTVTVAVADRIPDILADLGSIKTPIDPEDLDLTLYLVVAVAAVSCVFLAFVIVLLVLRLRRWHKSRLLQAEGSRLAGVPASHFVGVDGVRAFLQTYSHEVSLTADSRKSHLIFPQPNYADTLLSEESCEKSEPLLMSDKVDANKEERRVQVSFLFRSpecifications
| protocadherin gamma subfamily A, 5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human PCDHGA5 protein. | |
| PCDHGA5 | |
| CDH-GAMMA-A5/ME3/MGC142020/PCDH-GAMMA-A5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_032054.1 | |
| PCDHGA5 | |
| PCDHGA5 (NP_114443.1, 1 a.a. ∼ 813 a.a) full-length human protein. | |
| 100 μg | |
| Cell Adhesion | |
| 56110 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.